Product Details

SNP ID
rs150835153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99329920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGAAGCGGAGCGGGGCGGCGCGG[A/G]CGGTGGCGGGAAGCTCTTTTCACTC
Phenotype
MIM: 607802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNNM1 PubMed Links

Gene Details

Gene
CNNM1
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020348.2 822 Missense Mutation GAC,GGC D178G NP_065081.2
XM_005269692.3 822 Missense Mutation GAC,GGC D178G XP_005269749.2
XM_011539629.1 822 Missense Mutation GAC,GGC D178G XP_011537931.1
XM_011539630.1 822 Missense Mutation GAC,GGC D178G XP_011537932.1
XM_011539631.2 822 Missense Mutation GAC,GGC D178G XP_011537933.1

View Full Product Details