Product Details

SNP ID
rs151114097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:98251112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCGGTGCCCATCATACTTGCAG[C/T]GGCACTGCAGCATATTGTTGGAGAA
Phenotype
MIM: 607318
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOXL4 PubMed Links

Gene Details

Gene
LOXL4
Gene Name
lysyl oxidase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032211.6 2470 Missense Mutation CAC,CGC H718R NP_115587.6
XM_005270216.1 2470 Missense Mutation CAC,CGC H719R XP_005270273.1
XM_006718020.1 2470 Missense Mutation CAC,CGC H719R XP_006718083.1
Gene
R3HCC1L
Gene Name
R3H domain and coiled-coil containing 1 like
There are no transcripts associated with this gene.

View Full Product Details