Product Details

SNP ID
rs111438586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63946106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATACCTGTGTGTTGGAAGTTGGC[A/G]CCAGCAGTTGGCCCAGGTGTTTGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAA40 PubMed Links

Gene Details

Gene
NAA40
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300800.1 Intron NP_001287729.1
NM_024771.3 Intron NP_079047.2
XM_006718689.3 Intron XP_006718752.1
XM_011545254.2 Intron XP_011543556.1
XM_011545255.2 Intron XP_011543557.1

View Full Product Details