Product Details

SNP ID
rs111941251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56360697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTGGTCTCCCTCACATACCTC[C/T]ATGGCTTTTCTACAGCTATTGTGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8J1 PubMed Links

Gene Details

Gene
OR8J1
Gene Name
olfactory receptor family 8 subfamily J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005205.2 Intron NP_001005205.2

View Full Product Details