Product Details

SNP ID
rs112568150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66295221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTTTGGCGCCAGCTCGCCCCC[C/G]CCGGGGGCCGTGCCCAGCGGGCCCC
Phenotype
MIM: 611484
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM151A PubMed Links

Gene Details

Gene
TMEM151A
Gene Name
transmembrane protein 151A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153266.3 1087 Silent Mutation CCC,CCG P325P NP_694998.1
Gene
YIF1A
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
There are no transcripts associated with this gene.

View Full Product Details