Product Details

SNP ID
rs113936495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64853941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGTGCCTGGTGTTCCTGTCCAT[C/G]CAGCCTCAGCCTAGCAGCTGCCAAA
Phenotype
MIM: 613991 MIM: 605888
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC42BPG PubMed Links

Gene Details

Gene
CDC42BPG
Gene Name
CDC42 binding protein kinase gamma
There are no transcripts associated with this gene.

Gene
EHD1
Gene Name
EH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282444.1 2122 UTR 3 NP_001269373.1
NM_001282445.1 2122 UTR 3 NP_001269374.1
NM_006795.3 2122 UTR 3 NP_006786.2
XM_011544739.1 2122 UTR 3 XP_011543041.1

View Full Product Details