Product Details

SNP ID
rs115046286
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63574889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAACCAAACCCCAAACTCTCTAG[C/T]AAAACAAGACCCCCTTGATGTATAA
Phenotype
MIM: 613866 MIM: 613867
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HRASLS2 PubMed Links
Additional Information
For this assay, SNP(s) [rs145239735] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HRASLS2
Gene Name
HRAS like suppressor 2
There are no transcripts associated with this gene.

Gene
PLA2G16
Gene Name
phospholipase A2 group XVI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128203.1 870 UTR 3 NP_001121675.1
NM_007069.3 870 UTR 3 NP_009000.2
XM_006718426.1 870 UTR 3 XP_006718489.1
XM_011544741.1 870 UTR 3 XP_011543043.1

View Full Product Details