Product Details

SNP ID
rs115772206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68980330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGAGGCCAGGACCACCACGAA[C/T]AGCCGTGTGGGCTGCCGCCGCCACT
Phenotype
MIM: 607231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRGPRD PubMed Links

Gene Details

Gene
MRGPRD
Gene Name
MAS related GPR family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198923.2 657 Silent Mutation CTA,CTG L219L NP_944605.2

View Full Product Details