Product Details

SNP ID
rs117326881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68710332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAACAAATGAATGGAATCTAATA[A/G]ATAGTAAAACTATGACTCAACTTTG
Phenotype
MIM: 604374
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TESMIN PubMed Links

Gene Details

Gene
TESMIN
Gene Name
testis expressed metallothionein like protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039656.1 Intron NP_001034745.1
NM_004923.3 Intron NP_004914.2
XM_011545402.1 Intron XP_011543704.1
XM_011545403.2 Intron XP_011543705.1
XM_017018588.1 Intron XP_016874077.1

View Full Product Details