Product Details

SNP ID
rs137853935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67365138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGAAGAGGAGAAGGTCAGAAGTC[G/A]GAAGCCATGAGCCCCCAGGTGCAGG
Phenotype
MIM: 607672
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CLCF1 PubMed Links

Gene Details

Gene
CLCF1
Gene Name
cardiotrophin-like cytokine factor 1
There are no transcripts associated with this gene.

Gene
LOC100130987
Gene Name
uncharacterized LOC100130987
There are no transcripts associated with this gene.

View Full Product Details