Product Details

SNP ID
rs137879055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67367464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCTACGCTGGATACTCACATAG[A/G]TCCCAGCCAAGCTGCGGAGTTGGTG
Phenotype
MIM: 607672
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCF1 PubMed Links

Gene Details

Gene
CLCF1
Gene Name
cardiotrophin-like cytokine factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166212.1 294 Missense Mutation ACC,ATC T50I NP_001159684.1
NM_013246.2 294 Missense Mutation ACC,ATC T60I NP_037378.1
Gene
LOC100130987
Gene Name
uncharacterized LOC100130987
There are no transcripts associated with this gene.

View Full Product Details