Product Details

SNP ID
rs137903095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111354519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACGGGAAATAGGTGAGGGGTGC[C/T]TGGTGCTCTGGGCCCTCCAGCGGGG
Phenotype
MIM: 601206
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4491 PubMed Links

Gene Details

Gene
MIR4491
Gene Name
microRNA 4491
There are no transcripts associated with this gene.

Gene
POU2AF1
Gene Name
POU class 2 associating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006235.2 2469 Silent Mutation CAA,CAG Q171Q NP_006226.2
XM_005271593.1 2469 Silent Mutation CAA,CAG Q218Q XP_005271650.1
XM_005271594.3 2469 Silent Mutation CAA,CAG Q218Q XP_005271651.1
XM_006718859.1 2469 Silent Mutation CAA,CAG Q191Q XP_006718922.1
XM_006718860.3 2469 UTR 3 XP_006718923.1
XM_017017932.1 2469 UTR 3 XP_016873421.1

View Full Product Details