Product Details
- SNP ID
-
rs137992820
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:58422556 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAAGTGAGAGGCACAGGTGGACA[A/G]TGCTTTTTGGTGTCCCTTAGCTGAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR5B2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10466659] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR5B2
- Gene Name
- olfactory receptor family 5 subfamily B member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005566.2 |
758 |
Silent Mutation |
CTG,TTG |
L236L |
NP_001005566.1 |
View Full Product Details