Product Details

SNP ID
rs138308723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125789450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGACAGACCGCTGTAGAAGAGG[C/T]TTTGGTGTCTGTACTGCTCAGAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PATE3 PubMed Links

Gene Details

Gene
PATE3
Gene Name
prostate and testis expressed 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129883.3 3455 Silent Mutation GGC,GGT G38G NP_001123355.3
XM_017017054.1 3455 Silent Mutation GGC,GGT G26G XP_016872543.1

View Full Product Details