Product Details

SNP ID
rs138343311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:86250302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATTTACTGCTTACAGGATGAC[A/G]CTGTCAGTATAGAAAGTGGTACAAA
Phenotype
MIM: 605984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EED PubMed Links

Gene Details

Gene
EED
Gene Name
embryonic ectoderm development
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308007.1 807 Missense Mutation ACT,GCT T41A NP_001294936.1
NM_003797.4 807 Missense Mutation ACT,GCT T41A NP_003788.2
XM_005274373.2 807 Missense Mutation ACT,GCT T41A XP_005274430.1
XM_011545330.1 807 Missense Mutation ACT,GCT T41A XP_011543632.1
XM_011545331.1 807 Missense Mutation ACT,GCT T41A XP_011543633.1
XM_017018512.1 807 Missense Mutation ACT,GCT T41A XP_016874001.1
XM_017018513.1 807 Missense Mutation ACT,GCT T41A XP_016874002.1

View Full Product Details