Product Details

SNP ID
rs138824010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119094999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGCCTGGGTGGCCTTCTTGCC[G/T]CCCGAGGGCGCCTTCGGCCCCACGG
Phenotype
MIM: 191350 MIM: 601772 MIM: 609806
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DPAGT1 PubMed Links

Gene Details

Gene
DPAGT1
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
There are no transcripts associated with this gene.

Gene
H2AFX
Gene Name
H2A histone family member X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002105.2 469 Silent Mutation NP_002096.1
Gene
HMBS
Gene Name
hydroxymethylbilane synthase
There are no transcripts associated with this gene.

View Full Product Details