Product Details

SNP ID
rs139034795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:120341668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTTGCATCATGGTTGTCTGAC[A/G]TTATTAGAAATGTTTCTTATATACT
Phenotype
MIM: 604763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF12 PubMed Links

Gene Details

Gene
ARHGEF12
Gene Name
Rho guanine nucleotide exchange factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198665.1 Intron NP_001185594.1
NM_001301084.1 Intron NP_001288013.1
NM_015313.2 Intron NP_056128.1
XM_006718805.3 Intron XP_006718868.1
XM_011542720.2 Intron XP_011541022.1
XM_017017420.1 Intron XP_016872909.1
XM_017017421.1 Intron XP_016872910.1
XM_017017422.1 Intron XP_016872911.1
Gene
TMEM136
Gene Name
transmembrane protein 136
There are no transcripts associated with this gene.

View Full Product Details