Product Details
- SNP ID
-
rs139198011
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:3359334 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTAATGTCTGAGAACCACATGAA[C/G]ACTCTGCCACATTCTTCACACTTGT
- Phenotype
-
MIM: 602187
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF195
PubMed Links
Gene Details
- Gene
- ZNF195
- Gene Name
- zinc finger protein 195
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130519.2 |
2193 |
Silent Mutation |
GTC,GTG |
V535V |
NP_001123991.1 |
NM_001130520.2 |
2193 |
Silent Mutation |
GTC,GTG |
V558V |
NP_001123992.1 |
NM_001242841.1 |
2193 |
Silent Mutation |
GTC,GTG |
V539V |
NP_001229770.1 |
NM_001242842.1 |
2193 |
Silent Mutation |
GTC,GTG |
V513V |
NP_001229771.1 |
NM_001242843.1 |
2193 |
Silent Mutation |
GTC,GTG |
V490V |
NP_001229772.1 |
NM_001256823.1 |
2193 |
UTR 3 |
|
|
NP_001243752.1 |
NM_001256824.1 |
2193 |
Silent Mutation |
GTC,GTG |
V521V |
NP_001243753.1 |
NM_001256825.1 |
2193 |
Silent Mutation |
GTC,GTG |
V490V |
NP_001243754.1 |
NM_007152.4 |
2193 |
Silent Mutation |
GTC,GTG |
V486V |
NP_009083.2 |
XM_006718306.1 |
2193 |
Silent Mutation |
GTC,GTG |
V501V |
XP_006718369.1 |
XM_011520350.2 |
2193 |
Silent Mutation |
GTC,GTG |
V562V |
XP_011518652.1 |
XM_011520351.2 |
2193 |
Silent Mutation |
GTC,GTG |
V562V |
XP_011518653.1 |
XM_011520352.2 |
2193 |
Silent Mutation |
GTC,GTG |
V524V |
XP_011518654.1 |
XM_011520354.2 |
2193 |
Intron |
|
|
XP_011518656.2 |
XM_017018260.1 |
2193 |
Silent Mutation |
GTC,GTG |
V573V |
XP_016873749.1 |
XM_017018261.1 |
2193 |
Silent Mutation |
GTC,GTG |
V562V |
XP_016873750.1 |
XM_017018262.1 |
2193 |
Silent Mutation |
GTC,GTG |
V550V |
XP_016873751.1 |
XM_017018263.1 |
2193 |
Silent Mutation |
GTC,GTG |
V539V |
XP_016873752.1 |
XM_017018264.1 |
2193 |
Silent Mutation |
GTC,GTG |
V418V |
XP_016873753.1 |
XM_017018265.1 |
2193 |
Silent Mutation |
GTC,GTG |
V459V |
XP_016873754.1 |
XM_017018266.1 |
2193 |
Silent Mutation |
GTC,GTG |
V459V |
XP_016873755.1 |
XM_017018267.1 |
2193 |
Silent Mutation |
GTC,GTG |
V482V |
XP_016873756.1 |
View Full Product Details