Product Details
- SNP ID
-
hCV161096660
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:95026525 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTTTTCCATGGACCCCTTTGTGC[A/G]CATTGTGCAACCCGAGAGTTATGAG
- Phenotype
-
MIM: 616581
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KDM4E
PubMed Links
Gene Details
- Gene
- KDM4E
- Gene Name
- lysine demethylase 4E
There are no transcripts associated with this gene.
View Full Product Details