Product Details

SNP ID
rs139804672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7796499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTATAGGAGACAGCAATGAGAAA[A/G]CCAGCTATGTAAACTACTAAGAGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5P2 PubMed Links

Gene Details

Gene
OR5P2
Gene Name
olfactory receptor family 5 subfamily P member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153444.1 444 Silent Mutation GGC,GGT G148G NP_703145.1

View Full Product Details