Product Details

SNP ID
rs139806126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65965387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGACCCCTGGCTGGACGACAC[A/T]GCAGCCTGGATCGAGAGGAGCCGGC
Phenotype
MIM: 605941
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SART1 PubMed Links

Gene Details

Gene
SART1
Gene Name
squamous cell carcinoma antigen recognized by T-cells 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005146.4 721 Silent Mutation ACA,ACT T200T NP_005137.1
XM_011545344.1 721 Silent Mutation ACA,ACT T102T XP_011543646.1
XM_011545345.2 721 Silent Mutation ACA,ACT T42T XP_011543647.1
Gene
TSGA10IP
Gene Name
testis specific 10 interacting protein
There are no transcripts associated with this gene.

View Full Product Details