Product Details

SNP ID
rs139818945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68032324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGCCTCCACAGCAGTGCAGTG[A/G]CAGCCACCTACAGTGAGTACCTGGG
Phenotype
MIM: 600466 MIM: 602141 MIM: 604592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH3B1 PubMed Links

Gene Details

Gene
ALDH3B1
Gene Name
aldehyde dehydrogenase 3 family member B1
There are no transcripts associated with this gene.

Gene
MIR4691
Gene Name
microRNA 4691
There are no transcripts associated with this gene.

Gene
MIR7113
Gene Name
microRNA 7113
There are no transcripts associated with this gene.

Gene
NDUFS8
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002496.3 311 Missense Mutation ACA,GCA T33A NP_002487.1
XM_005274013.1 311 Missense Mutation ACA,GCA T33A XP_005274070.1
XM_005274014.2 311 Missense Mutation ACA,GCA T33A XP_005274071.1
XM_011545053.2 311 Missense Mutation ACA,GCA T33A XP_011543355.1
Gene
TCIRG1
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
There are no transcripts associated with this gene.

View Full Product Details