Product Details

SNP ID
rs139859991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4682022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAAGGCCTTGGCCCGCTCTGA[A/C]TTGGAAGGCAGTTGCAGAACCGTTC
Phenotype
MIM: 611268
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR51E2 PubMed Links

Gene Details

Gene
OR51E2
Gene Name
olfactory receptor family 51 subfamily E member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030774.3 934 Missense Mutation AAG,AAT K230N NP_110401.1

View Full Product Details