Product Details

SNP ID
rs139938503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55638681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCACCCATTTTTTTGGAGGCAT[A/G]GAGATCTTCATTCTCACAGGGATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4P4 PubMed Links

Gene Details

Gene
OR4P4
Gene Name
olfactory receptor family 4 subfamily P member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004124.2 324 Missense Mutation ATA,ATG I108M NP_001004124.1

View Full Product Details