Product Details
- SNP ID
-
rs140107613
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:63914032 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGGTCCGGGCTTCCTGACACTGC[C/T]GTGAGGCCTTCAGGGCTCAGGTACA
- Phenotype
-
MIM: 600526
MIM: 616019
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MARK2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10897461] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MARK2
- Gene Name
- microtubule affinity regulating kinase 2
There are no transcripts associated with this gene.
- Gene
- RCOR2
- Gene Name
- REST corepressor 2
View Full Product Details