Product Details

SNP ID
rs140187145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113386874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGACTTACTCAGCTGTAGGCA[A/G]ATAGTGGCTGACTCTGCACTCCGTG
Phenotype
MIM: 608774 MIM: 610732
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKK1 PubMed Links

Gene Details

Gene
ANKK1
Gene Name
ankyrin repeat and kinase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178510.1 861 Intron NP_848605.1
XM_011542736.2 861 UTR 5 XP_011541038.1
XM_011542737.2 861 UTR 5 XP_011541039.1
XM_011542738.2 861 Intron XP_011541040.1
XM_017017475.1 861 UTR 5 XP_016872964.1
Gene
TTC12
Gene Name
tetratricopeptide repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318533.1 861 Intron NP_001305462.1
NM_017868.3 861 Intron NP_060338.3
XM_005271604.3 861 Intron XP_005271661.1
XM_011542890.2 861 Intron XP_011541192.1
XM_017017943.1 861 Intron XP_016873432.1
XM_017017944.1 861 Intron XP_016873433.1
XM_017017945.1 861 Intron XP_016873434.1
XM_017017946.1 861 Intron XP_016873435.1
XM_017017947.1 861 Intron XP_016873436.1
XM_017017948.1 861 Intron XP_016873437.1
XM_017017949.1 861 Intron XP_016873438.1
XM_017017950.1 861 Intron XP_016873439.1

View Full Product Details