Product Details

SNP ID
rs140455461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61435730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGAGCTGCTACAAGTCTCCCA[A/G]CTGTCGGTGGCCTCCAGACTTTTAG
Phenotype
MIM: 613019
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPSF7 PubMed Links

Gene Details

Gene
CPSF7
Gene Name
cleavage and polyadenylation specific factor 7
There are no transcripts associated with this gene.

Gene
SDHAF2
Gene Name
succinate dehydrogenase complex assembly factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017841.2 Intron NP_060311.1

View Full Product Details