Product Details

SNP ID
rs140460244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56346629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGATGTCCATACTCTGTTCTGA[C/G]ACAAATGAATTAGAATTAATAATTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR8K1 PubMed Links

Gene Details

Gene
OR8K1
Gene Name
olfactory receptor family 8 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002907.1 591 Missense Mutation GAC,GAG D197E NP_001002907.1

View Full Product Details