Product Details

SNP ID
rs140715783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46859299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTTGATCTTCTCCTTGGTGTAG[G/T]TATGGTCAGGCCCTCCCTAGGGTGG
Phenotype
MIM: 604270
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRP4 PubMed Links

Gene Details

Gene
LRP4
Gene Name
LDL receptor related protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002334.3 4715 Missense Mutation NP_002325.2
XM_011520103.2 4715 Missense Mutation XP_011518405.1
XM_011520104.2 4715 Missense Mutation XP_011518406.1
XM_017017734.1 4715 UTR 3 XP_016873223.1
Gene
LRP4-AS1
Gene Name
LRP4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details