Product Details

SNP ID
rs141037265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123940084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGTGGGGGGCTTCATCCACTC[C/T]ATAGTACAGATTGCATTGACTATCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4D5 PubMed Links

Gene Details

Gene
OR4D5
Gene Name
olfactory receptor family 4 subfamily D member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001965.1 468 Silent Mutation TCC,TCT S156S NP_001001965.1
Gene
OR6T1
Gene Name
olfactory receptor family 6 subfamily T member 1
There are no transcripts associated with this gene.

View Full Product Details