Product Details

SNP ID
rs141073907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72220940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGCTGCACAAGGACACCTCCC[A/G]CCTGTACAACTTTAACTGGGACCAC
Phenotype
MIM: 136425 MIM: 600829
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLR2 PubMed Links

Gene Details

Gene
FOLR2
Gene Name
folate receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000803.4 378 Missense Mutation CAC,CGC H74R NP_000794.3
NM_001113534.1 378 Missense Mutation CAC,CGC H74R NP_001107006.1
NM_001113535.1 378 Missense Mutation CAC,CGC H74R NP_001107007.1
NM_001113536.1 378 Missense Mutation CAC,CGC H74R NP_001107008.1
XM_005273856.3 378 Missense Mutation CAC,CGC H83R XP_005273913.1
Gene
INPPL1
Gene Name
inositol polyphosphate phosphatase like 1
There are no transcripts associated with this gene.

View Full Product Details