Product Details

SNP ID
rs141141857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:19055691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGGAACCACTGAATGCCAAAGG[C/G]CAGGCCGCAGAGGAGGAACACCAGC
Phenotype
MIM: 607228
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRGPRX2 PubMed Links

Gene Details

Gene
MRGPRX2
Gene Name
MAS related GPR family member X2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303615.1 800 Missense Mutation CCC,GCC P238A NP_001290544.1
NM_054030.3 800 Missense Mutation CCC,GCC P238A NP_473371.1

View Full Product Details