Product Details

SNP ID
rs142147233
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60337890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATACCAACCAGACTTCTTCTTTC[G/T]TATGATTATCATTCACCTTACACCA
Phenotype
MIM: 608402
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MS4A6E PubMed Links
Additional Information
For this assay, SNP(s) [rs2289613] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MS4A6E
Gene Name
membrane spanning 4-domains A6E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139249.2 Intron NP_640342.1

View Full Product Details