Product Details

SNP ID
rs142248522
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119342601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGGGCAGGCTTCTCACCTGGG[G/T]GTGGGAACAAGGGGCCGCTGCAGTT
Phenotype
MIM: 608752 MIM: 606227 MIM: 606130
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
C1QTNF5 PubMed Links
Additional Information
For this assay, SNP(s) [rs11217241] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C1QTNF5
Gene Name
C1q and tumor necrosis factor related protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278431.1 1542 Intron NP_001265360.1
NM_015645.4 1542 UTR 5 NP_056460.1
Gene
MFRP
Gene Name
membrane frizzled-related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031433.3 1542 Missense Mutation CAC,CCC H461P NP_113621.1
Gene
RNF26
Gene Name
ring finger protein 26
There are no transcripts associated with this gene.

View Full Product Details