Product Details
- SNP ID
-
rs142458921
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:67451440 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATGCGAGGATGGGGTTGGGGAAG[C/T]TTCATCACAGGGACTGGGCACAGAA
- Phenotype
-
MIM: 608965
MIM: 609849
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CABP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs949252] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CABP4
- Gene Name
- calcium binding protein 4
- Gene
- CORO1B
- Gene Name
- coronin 1B
There are no transcripts associated with this gene.
- Gene
- GPR152
- Gene Name
- G protein-coupled receptor 152
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_206997.1 |
1290 |
Missense Mutation |
ACT,GCT |
T429A |
NP_996880.1 |
View Full Product Details