Product Details

SNP ID
rs142931474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:12286901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTGATGTGTGAACAAATGAAA[A/T]GTTTGGTAGAGTGTGTTGTGGGGAC
Phenotype
MIM: 608881 MIM: 612355
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MICAL2 PubMed Links

Gene Details

Gene
MICAL2
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 2
There are no transcripts associated with this gene.

Gene
MICALCL
Gene Name
MICAL C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032867.2 2 UTR 5 NP_116256.2

View Full Product Details