Product Details

SNP ID
rs143059012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57600043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGCCGACAACCAACTCAACAAC[A/C]AATTCAGCCACCAAAATAACAGCTA
Phenotype
MIM: 606860
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SERPING1 PubMed Links

Gene Details

Gene
SERPING1
Gene Name
serpin family G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000062.2 255 Silent Mutation ACA,ACC T72T NP_000053.2
NM_001032295.1 255 Silent Mutation ACA,ACC T72T NP_001027466.1

View Full Product Details