Product Details

SNP ID
rs143743236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5389726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAGTATCTACTTTGGAGCGTGT[A/C]AAATCCAGATGTTCTGCATCCACTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 350 Intron NP_001005567.2
Gene
OR51M1
Gene Name
olfactory receptor family 51 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004756.2 350 Missense Mutation AAA,CAA K110Q NP_001004756.2

View Full Product Details