Product Details

SNP ID
rs143767594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123884128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGCAGTATACATGATCCAGGTG[A/G]TCCTATAACTAGAGAAGTGCATGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM225 PubMed Links

Gene Details

Gene
TMEM225
Gene Name
transmembrane protein 225
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013743.2 680 Missense Mutation ACC,ATC T137I NP_001013765.2
XM_006718832.3 680 Missense Mutation ACC,ATC T87I XP_006718895.1
XM_011542802.2 680 Intron XP_011541104.1
XM_017017653.1 680 Intron XP_016873142.1

View Full Product Details