Product Details

SNP ID
rs144356023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5884504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTCCTGGCCATGTTGTCTATGA[C/T]TGATCTGGGTCTGTCCACATCCACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52E4 PubMed Links

Gene Details

Gene
OR52E4
Gene Name
olfactory receptor family 52 subfamily E member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005165.1 212 Missense Mutation ACT,ATT T71I NP_001005165.1

View Full Product Details