Product Details
- SNP ID
-
rs145837350
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5489050 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCATTCTATCTATGCTCTGGAGT[C/T]CTCAATTCTACTTGCCATGGCCTTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR51B5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs417425] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR51B5
- Gene Name
- olfactory receptor family 51 subfamily B member 5
- Gene
- OR52D1
- Gene Name
- olfactory receptor family 52 subfamily D member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005163.2 |
366 |
Missense Mutation |
TCC,TTC |
S115F |
NP_001005163.1 |
View Full Product Details