Product Details

SNP ID
rs146094810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2888277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAAATCCTGCAAGCGCGGGGTTG[A/G]GGGGGCGGGGGAGACCGCTGTCGGG
Phenotype
MIM: 600856 MIM: 603240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDKN1C PubMed Links

Gene Details

Gene
CDKN1C
Gene Name
cyclin dependent kinase inhibitor 1C
There are no transcripts associated with this gene.

Gene
SLC22A18AS
Gene Name
solute carrier family 22 member 18 antisense
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302862.1 2788 Missense Mutation NP_001289791.1
NM_007105.3 2788 Missense Mutation NP_009036.2
XM_017017832.1 2788 Missense Mutation XP_016873321.1
XM_017017833.1 2788 Missense Mutation XP_016873322.1
XM_017017834.1 2788 Missense Mutation XP_016873323.1
XM_017017835.1 2788 Missense Mutation XP_016873324.1
XM_017017836.1 2788 Missense Mutation XP_016873325.1

View Full Product Details