Product Details

SNP ID
rs146176077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:122664357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGAGTGGGGAGAAAAAACCAC[A/G]CTCAAGAATGGATCCTCTCTGCAGG
Phenotype
MIM: 609201
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBASH3B PubMed Links

Gene Details

Gene
UBASH3B
Gene Name
ubiquitin associated and SH3 domain containing B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032873.4 Intron NP_116262.2
XM_005271712.3 Intron XP_005271769.1
XM_011543041.2 Intron XP_011541343.1
XM_011543042.1 Intron XP_011541344.1

View Full Product Details