Product Details

SNP ID
rs146427121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5777861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAATGCTGGAACATAGGTGAT[A/G]ATGATGGCAGATATATGGGCAGTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52N1 PubMed Links

Gene Details

Gene
OR52N1
Gene Name
olfactory receptor family 52 subfamily N member 1
There are no transcripts associated with this gene.

Gene
OR52N5
Gene Name
olfactory receptor family 52 subfamily N member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001922.2 807 Silent Mutation ATC,ATT I258I NP_001001922.2

View Full Product Details