Product Details

SNP ID
rs146523706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5856823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCCTGACTCCATAGATTACAG[A/G]ATTGAGGGCTGGTGGGACAACCACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52E8 PubMed Links

Gene Details

Gene
OR52E8
Gene Name
olfactory receptor family 52 subfamily E member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005168.1 880 Missense Mutation CCT,TCT P294S NP_001005168.1

View Full Product Details