Product Details

SNP ID
rs146962720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18934565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATAAGGCGGCCGCTGAGGAAGA[A/G]GAAGTCTGCTGCGGCCAAGTTGAGG
Phenotype
MIM: 607227
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRGPRX1 PubMed Links

Gene Details

Gene
MRGPRX1
Gene Name
MAS related GPR family member X1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147199.3 Intron NP_671732.3

View Full Product Details