Product Details

SNP ID
rs147065932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1277174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGATGGCTTTCAGGTCCTCCTC[A/G]CTACAGCGGGGCTGGGCGTTCACGG
Phenotype
MIM: 606277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TOLLIP PubMed Links

Gene Details

Gene
TOLLIP
Gene Name
toll interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318512.1 854 Silent Mutation AGC,AGT S180S NP_001305441.1
NM_001318514.1 854 Silent Mutation AGC,AGT S161S NP_001305443.1
NM_001318515.1 854 UTR 3 NP_001305444.1
NM_001318516.1 854 Silent Mutation AGC,AGT S169S NP_001305445.1
NM_019009.3 854 Silent Mutation AGC,AGT S230S NP_061882.2
XM_017017931.1 854 Silent Mutation AGC,AGT S104S XP_016873420.1

View Full Product Details