Product Details

SNP ID
rs147336850
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18609865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAAAAATAAAAGCAGCAGCTAA[C/T]GCCTCTTCAGCTTCTTGGCCCTTCG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SPTY2D1 PubMed Links

Gene Details

Gene
SPTY2D1
Gene Name
SPT2 chromatin protein domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194285.2 2128 Missense Mutation NP_919261.2
XM_011519919.2 2128 Missense Mutation XP_011518221.1
Gene
SPTY2D1-AS1
Gene Name
SPTY2D1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details