Product Details

SNP ID
rs147455935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68032301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTGCAGGACCTCCTGGTGGCC[A/G]GAGCCTCCACAGCAGTGCAGTGGCA
Phenotype
MIM: 600466 MIM: 602141 MIM: 604592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH3B1 PubMed Links

Gene Details

Gene
ALDH3B1
Gene Name
aldehyde dehydrogenase 3 family member B1
There are no transcripts associated with this gene.

Gene
MIR4691
Gene Name
microRNA 4691
There are no transcripts associated with this gene.

Gene
MIR7113
Gene Name
microRNA 7113
There are no transcripts associated with this gene.

Gene
NDUFS8
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002496.3 288 Missense Mutation CAG,CGG Q25R NP_002487.1
XM_005274013.1 288 Missense Mutation CAG,CGG Q25R XP_005274070.1
XM_005274014.2 288 Missense Mutation CAG,CGG Q25R XP_005274071.1
XM_011545053.2 288 Missense Mutation CAG,CGG Q25R XP_011543355.1
Gene
TCIRG1
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
There are no transcripts associated with this gene.

View Full Product Details