Product Details

SNP ID
rs147460978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:197595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACACGCCCACCAAGCTGCGTGC[A/G]CCGGCCTACAGCTTCCGTGGGGCCC
Phenotype
MIM: 615417 MIM: 608356 MIM: 609146 MIM: 610176
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BET1L PubMed Links

Gene Details

Gene
BET1L
Gene Name
Bet1 golgi vesicular membrane trafficking protein like
There are no transcripts associated with this gene.

Gene
LOC388572
Gene Name
uncharacterized LOC388572
There are no transcripts associated with this gene.

Gene
ODF3
Gene Name
outer dense fiber of sperm tails 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286136.1 438 Silent Mutation GCA,GCG A48A NP_001273065.1
NM_053280.4 438 Silent Mutation GCA,GCG A48A NP_444510.2
Gene
RIC8A
Gene Name
RIC8 guanine nucleotide exchange factor A
There are no transcripts associated with this gene.

Gene
SCGB1C1
Gene Name
secretoglobin family 1C member 1
There are no transcripts associated with this gene.

View Full Product Details